3'-Bromoacetophenone, 97%
3'-Bromoacetophenone, 97%
3'-Bromoacetophenone, 97%
3'-Bromoacetophenone, 97%
Thermo Scientific Chemicals
3'-Bromoacetophenone, 97%
CAS: 2142-63-4 | C8H7BrO | 199.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA106580050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3'-Bromoacetophenone
CAS2142-63-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0