Bromoacetic acid, 98+%
Bromoacetic acid, 98+%
Bromoacetic acid, 98+%
Bromoacetic acid, 98+%
Thermo Scientific Chemicals
Bromoacetic acid, 98+%
CAS: 79-08-3 | C2H3BrO2 | 138.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA106572500
Price (MYR)
413.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
413.00
EA
Add to cart
Specifications
Chemical Name or MaterialBromoacetic acid
CAS79-08-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air