1,2-Bis(4-pyridyl)ethane, 97%
1,2-Bis(4-pyridyl)ethane, 97%
1,2-Bis(4-pyridyl)ethane, 97%
1,2-Bis(4-pyridyl)ethane, 97%
Thermo Scientific Chemicals
1,2-Bis(4-pyridyl)ethane, 97%
CAS: 4916-57-8 | C12H12N2 | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA106470100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Bis(4-pyridyl)ethane
CAS4916-57-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0