Methylthymol blue, tetrasodium salt, pure, water soluble
CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol
Have Questions?
Catalog Number
Quantity
FSA106370050
5 g
Catalog number FSA106370050
Price (MYR)
443.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
443.00
EA
Add to cart
Specifications
Absorbance130 min. min. (at lambda max, in water)
Chemical Name or MaterialMethylthymol Blue, tetrasodium salt
CAS1945-77-3
EINECS Number217-743-3
TSCATSCA