Biphenyl, 99%
Biphenyl, 99%
Biphenyl, 99%
Thermo Scientific Chemicals
Biphenyl, 99%
CAS: 92-52-4 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA106252500
Price (MYR)
251.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
251.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBiphenyl
CAS92-52-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0