Bibenzyl, 99%
Bibenzyl, 99%
Bibenzyl, 99%
Bibenzyl, 99%
CAS: 103-29-7 | C14H14 | 182.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA106235000
Price (MYR)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBibenzyl
Name Note99%
CAS103-29-7
CAS Min %98.5
CAS Max %100.0