Benzyl bromide, 98%
Benzyl bromide, 98%
Benzyl bromide, 98%
Thermo Scientific Chemicals
Benzyl bromide, 98%
CAS: 100-39-0 | C7H7Br | 171.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA105875000
Price (MYR)
1,735.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,735.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyl bromide
Name Note98%
CAS100-39-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
Alkylation reagent
RUO – Research Use Only