Benzylamine, 99%, pure
Benzylamine, 99%, pure
Benzylamine, 99%, pure
Benzylamine, 99%, pure
Thermo Scientific Chemicals
Benzylamine, 99%, pure
CAS: 100-46-9 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA105855000
Price (MYR)
491.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
491.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzylamine
Name Note99%
CAS100-46-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger