Benzylacetone, 99%
Benzylacetone, 99%
Benzylacetone, 99%
Thermo Scientific Chemicals
Benzylacetone, 99%
CAS: 2550-26-7 | C10H12O | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number FSA105832500
Price (MYR)
420.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
420.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzylacetone
Name Note98%
CAS2550-26-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning