1,4-Benzoquinone, 98+%
1,4-Benzoquinone, 98+%
1,4-Benzoquinone, 98+%
1,4-Benzoquinone, 98+%
Thermo Scientific Chemicals
1,4-Benzoquinone, 98+%
1, 4-Benzoquinone, 99%, C6H4O2, CAS Number-106-51-4, 1,4-benzoquine, 1,4-benzoquinone, 2,5-cyclohexadiene-1,4-dione, benzoquinone, chinone, cyclohexadienedione, para-quinone, p-benzoquinone, p-quinone, quinone, 1kg, 98.5% min. (Iodometry), 07,609, 100.0, CHEBI:16509, Yellow to Green, 1.3100g/mL | CAS: 106-51-4 | C6H4O2 | 108.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA105635000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Benzoquinone
CAS106-51-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0