Benzophenone, 99%, pure
Benzophenone, 99%, pure
Benzophenone, 99%, pure
Benzophenone, 99%, pure
Thermo Scientific Chemicals
Benzophenone, 99%, pure
CAS: 119-61-9 | C13H10O | 182.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA105565000
Price (MYR)
332.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
332.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzophenone
CAS119-61-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0