Benzonitrile, 99%, pure
Benzonitrile, 99%, pure
Benzonitrile, 99%, pure
Benzonitrile, 99%, pure
Thermo Scientific Chemicals
Benzonitrile, 99%, pure
CAS: 100-47-0 | C7H5N | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA105540010
Price (MYR)
533.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
533.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzonitrile
CAS100-47-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0