1,2,4-Benzenetricarboxylic anhydride, 97%
1,2,4-Benzenetricarboxylic anhydride, 97%
1,2,4-Benzenetricarboxylic anhydride, 97%
1,2,4-Benzenetricarboxylic anhydride, 97%
Thermo Scientific Chemicals
1,2,4-Benzenetricarboxylic anhydride, 97%
CAS: 552-30-7 | C9H4O5 | 192.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA105370010
Price (MYR)
567.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
567.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2, 4-Benzenetricarboxylic anhydride
Name Note97%
CAS552-30-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger