1,3,5-Benzenetricarboxylic acid, 98%
1,3,5-Benzenetricarboxylic acid, 98%
1,3,5-Benzenetricarboxylic acid, 98%
1,3,5-Benzenetricarboxylic acid, 98%
Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid, 98%
1, 3, 5-Benzenetricarboxylic acid, 98%, C9H6O6, CAS Number-554-95-0, 1,3,5-benzene tricarboxylic acid, 1,3,5-benzenetricarboxylic acid, 1,3,5-tricarboxybenzene, 5-carboxyisophthalic acid, chembl77562, ou36oo5mtn, trimesic acid, trimesinic acid, trimesitinic acid, unii-ou36oo5mtn, 500g | CAS: 554-95-0 | C9H6O6 | 210.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
50 g
Packaging:
View all
Plastic bottle
Catalog number FSA105355000
Price (MYR)
1,731.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,731.00
EA
Add to cart
Specifications
Chemical Name or Material1, 3, 5-Benzenetricarboxylic acid
CAS554-95-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0