1,2-Benzanthracene, 99%
1,2-Benzanthracene, 99%
1,2-Benzanthracene, 99%
1,2-Benzanthracene, 99%
Thermo Scientific Chemicals
1,2-Benzanthracene, 99%
CAS: 56-55-3 | C18H12 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA105252500
Price (MYR)
424.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
424.00
EA
Add to cart
Specifications
Chemical Name or Material1, 2-Benzanthracene
Name Note99%
CAS56-55-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger