DL-Aspartic acid, 99+%
DL-Aspartic acid, 99+%
DL-Aspartic acid, 99+%
DL-Aspartic acid, 99+%
Thermo Scientific Chemicals
DL-Aspartic acid, 99+%
CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA105032500250 gPlastic Bottle
Catalog number FSA105032500
Price (MYR)
360.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
360.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-Aspartic acid
Name Note99+%
Ammonium500ppm max.
CAS617-45-8
ColorYellow