Anthrone, ACS reagent
Anthrone, ACS reagent
Anthrone, ACS reagent
Anthrone, ACS reagent
Anthrone, ACS reagent
CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Plastic Bottle
Catalog number FSA104965000
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAnthrone
CAS90-44-8
Absorbance0.75 max. (425nm, 1cm cell) Passes Test, 0.045 max. (620nm, 1cm cell) Passes Test
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: