Anthraquinone, 98%
Anthraquinone, 98%
Anthraquinone, 98%
Anthraquinone, 98%
Thermo Scientific Chemicals
Anthraquinone, 98%
CAS: 84-65-1 | C14H8O2 | 208.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA104932500
Price (MYR)
414.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
414.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAnthraquinone
CAS84-65-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention
View more
Suitable for battery materials development.
RUO – Research Use Only