Anthranilic acid, 98+%
Anthranilic acid, 98+%
Thermo Scientific Chemicals
Anthranilic acid, 98+%
CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA104911000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAnthranilic acid
Melting Point144.0°C to 148.0°C
CAS118-92-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Causes serious eye damage.