Anthracene, 99%
Anthracene, 99%
Anthracene, 99%
Thermo Scientific Chemicals
Anthracene, 99%
CAS: 120-12-7 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA104865000
Price (MYR)
1,301.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,301.00
EA
Add to cart
Specifications
Chemical Name or MaterialAnthracene
Name Note99%
CAS120-12-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.