p-Anisaldehyde, 99+%
p-Anisaldehyde, 99+%
p-Anisaldehyde, 99+%
p-Anisaldehyde, 99+%
Thermo Scientific Chemicals
p-Anisaldehyde, 99+%
CAS: 123-11-5 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA104805000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialp-Anisaldehyde
Name Note99+%
CAS123-11-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement: