D(-)-Amygdalin hydrate, 95%
D(-)-Amygdalin hydrate, 95%
D(-)-Amygdalin hydrate, 95%
D(-)-Amygdalin hydrate, 95%
Thermo Scientific Chemicals
D(-)-Amygdalin hydrate, 95%
CAS: 29883-15-6 | C20H27NO11 | 457.43 g/mol
Have Questions?
Catalog NumberQuantity
FSA1047300505 g
Catalog number FSA104730050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD(-)-Amygdalin hydrate
Name Note95%
CAS29883-15-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: