4-Aminopyridine, 98%
4-Aminopyridine, 98%
4-Aminopyridine, 98%
4-Aminopyridine, 98%
Thermo Scientific Chemicals
4-Aminopyridine, 98%
CAS: 504-24-5 | C5H6N2 | 94.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA104571000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminopyridine
Name Note98%
CAS504-24-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger