(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
Thermo Scientific Chemicals
(S)-(+)-2-Amino-1-propanol, 98%
CAS: 2749-11-3 | C3H9NO | 75.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA104450010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(S)-(+)-2-Amino-1-propanol
CAS2749-11-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0