3-Amino-1,2-propanediol, 98%
3-Amino-1,2-propanediol, 98%
3-Amino-1,2-propanediol, 98%
3-Amino-1,2-propanediol, 98%
Thermo Scientific Chemicals
3-Amino-1,2-propanediol, 98%
CAS: 616-30-8 | C3H9NO2 | 91.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA104420250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Amino-1, 2-propanediol
Name Note98%
CAS616-30-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger