2-Amino-5-picoline, 99%
2-Amino-5-picoline, 99%
2-Amino-5-picoline, 99%
2-Amino-5-picoline, 99%
Thermo Scientific Chemicals
2-Amino-5-picoline, 99%
CAS: 1603-41-4 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA104390100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-5-picoline
Name Note99%
CAS1603-41-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger