3-Amino-2-naphthoic acid, 85%, tech.
3-Amino-2-naphthoic acid, 85%, tech.
3-Amino-2-naphthoic acid, 85%, tech.
3-Amino-2-naphthoic acid, 85%, tech.
Thermo Scientific Chemicals
3-Amino-2-naphthoic acid, 85%, tech.
CAS: 5959-52-4 | C11H9NO2 | 187.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA104140050
Price (MYR)
741.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
741.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Amino-2-naphthoic acid
Name Note85%
CAS5959-52-4
CAS Min %80.0
Health Hazard 1GHS Signal Word: Warning