2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
Thermo Scientific Chemicals
2-Amino-2-methyl-1-propanol, 99%
CAS: 124-68-5 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number FSA104065000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-2-methyl-1-propanol
CAS124-68-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0