2-Amino-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-3-methylbenzoic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA103970100
Price (MYR)
439.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
439.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Amino-3-methylbenzoic acid
CAS4389-45-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0