N-(2-Aminoethyl)pyrrolidine, 99%
N-(2-Aminoethyl)pyrrolidine, 99%
N-(2-Aminoethyl)pyrrolidine, 99%
N-(2-Aminoethyl)pyrrolidine, 99%
Thermo Scientific Chemicals
N-(2-Aminoethyl)pyrrolidine, 99%
CAS: 7154-73-6 | C6H14N2 | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA103700050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-(2-Aminoethyl)pyrrolidine
CAS7154-73-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0