2-Amino-5-chlorobenzophenone, 98%
2-Amino-5-chlorobenzophenone, 98%
2-Amino-5-chlorobenzophenone, 98%
2-Amino-5-chlorobenzophenone, 98%
Thermo Scientific Chemicals
2-Amino-5-chlorobenzophenone, 98%
CAS: 719-59-5 | C13H10ClNO | 231.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA103345000
Price (MYR)
1,461.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,461.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-5-chlorobenzophenone
CAS719-59-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0