6-Aminocaproic acid, 99+%
6-Aminocaproic acid, 99+%
6-Aminocaproic acid, 99+%
6-Aminocaproic acid, 99+%
Thermo Scientific Chemicals
6-Aminocaproic acid, 99+%
CAS: 60-32-2 | C6H13NO2 | 131.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA103305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material6-Aminocaproic acid
CAS60-32-2
Health Hazard 1GHS Signal Word: Warning
Melting Point204.0°C to 209.0°C