4-Aminobenzophenone, 98%
4-Aminobenzophenone, 98%
4-Aminobenzophenone, 98%
4-Aminobenzophenone, 98%
Thermo Scientific Chemicals
4-Aminobenzophenone, 98%
CAS: 1137-41-3 | C13H11NO | 197.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA103170100
Price (MYR)
381.00
BOT
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
381.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Aminobenzophenone
Name Note98%
CAS1137-41-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning