4-Aminoantipyrine, 98%
4-Aminoantipyrine, 98%
4-Aminoantipyrine, 98%
4-Aminoantipyrine, 98%
Thermo Scientific Chemicals
4-Aminoantipyrine, 98%
4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow | CAS: 83-07-8 | C11H13N3O | 203.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA103155000
Price (MYR)
928.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
928.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Aminoantipyrine
Name Note98%
CAS83-07-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning