4'-Aminoacetophenone, 99%
4'-Aminoacetophenone, 99%
4'-Aminoacetophenone, 99%
4'-Aminoacetophenone, 99%
Thermo Scientific Chemicals
4'-Aminoacetophenone, 99%
CAS: 99-92-3 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA103095000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4'-Aminoacetophenone
CAS99-92-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0