Allyl Glycidyl Ether, 99+%
Allyl Glycidyl Ether, 99+%
Allyl Glycidyl Ether, 99+%
Allyl Glycidyl Ether, 99+%
Thermo Scientific Chemicals
Allyl Glycidyl Ether, 99+%
CAS: 106-92-3 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA102941000
Price (MYR)
318.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAllyl glycidyl ether
Name Note99+%
CAS106-92-3
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger