Allyl bromide, 99%, stabilized
Allyl bromide, 99%, stabilized
Allyl bromide, 99%, stabilized
Allyl bromide, 99%, stabilized
Thermo Scientific Chemicals
Allyl bromide, 99%, stabilized
CAS: 106-95-6 | C3H5Br | 120.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Packaging:
Glass bottle
Catalog number FSA102905000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAllyl bromide
Name NoteStabilized
CAS106-95-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
View more
Alkylation reagent
RUO – Research Use Only