2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
2-Acetylpyridine, 98%
Thermo Scientific Chemicals
2-Acetylpyridine, 98%
CAS: 1122-62-9 | C7H7NO | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA102685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Acetylpyridine
CAS1122-62-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0