2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Acetamido-2-deoxy-alpha-D-glucopyranose, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA102561000
Price (MYR)
764.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
764.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Acetamido-2-deoxy-α-D-glucopyranose
Name Note99.5+%
CAS10036-64-3
CAS Min %99.0
CAS Max %100.0