Acetonylacetone, 97%
Acetonylacetone, 97%
Acetonylacetone, 97%
Acetonylacetone, 97%
Thermo Scientific Chemicals
Acetonylacetone, 97%
CAS: 110-13-4 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA102401000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonylacetone
Name Note97%
CAS110-13-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning