4-Acetamidophenol, 98%
4-Acetamidophenol, 98%
4-Acetamidophenol, 98%
4-Acetamidophenol, 98%
Thermo Scientific Chemicals
4-Acetamidophenol, 98%
CAS: 103-90-2 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA102332500
Price (MYR)
420.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
420.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Acetamidophenol
CAS103-90-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0