2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA102310250
Price (MYR)
226.00
G
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
226.00
G
Add to cart
Specifications
Chemical Name or Material2-Acetamidophenol
CAS614-80-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0