Acetal, 99%
Acetal, 99%
Acetal, 99%
Thermo Scientific Chemicals
Acetal, 99%
CAS: 105-57-7 | C6H14O2 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number FSA102222500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetal
Name Note99%
CAS105-57-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger