Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Acenaphthenequinone, 95%
Thermo Scientific Chemicals
Acenaphthenequinone, 95%
CAS: 82-86-0 | C12H6O2 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA102190250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcenaphthenequinone
CAS82-86-0
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0