Q705 Sonicator with Probe
Q705 Sonicator with Probe
Actual product may vary
Q705 Sonicator with Probe
Use this high powered sonicator for any application and sample volume. Fisherbrand™ Sonicators are ideal for cell disruption, nanoparticle dispersion, DNA shearing/ChIP and homogenization.
Have Questions?
Catalog number FQSFB705EUK-220
Price (MYR)
-
  • 700W
  • Touch screen control
  • 10 programmable memories
  • Programmable from 1 second to 99 hours
  • Overload protection
  • Temperature monitoring
  • Amplitude control from 1 to 100%
  • Processing volume 0.2mL to 1L
Specifications
Voltage230 V
Electrical Requirements230V, 50/60Hz
Hertz20 kHz
Wattage700 W
Dimensions (L x W x H)387 x 203 x 216 mm (15.25 x 8 x 8.5 in.)
ModelQ705
Unit SizeEA