Probes for Model 50 and 120 Sonic Dismembrator
Probes for Model 50 and 120 Sonic Dismembrator
Actual product may vary
Probes for Model 50 and 120 Sonic Dismembrator
Probes for Model 50 and 120 Sonic Dismembrator
Probes for Model 50 and 120 Sonic Dismembrator
Probes for Model 50 and 120 Sonic Dismembrator
Image shows (left to right): 4423, 4422, 4435.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberAmplitudeDiameter (English)Diameter (Metric)Volume Range (Metric)
FQSFB4435120 μm maximum0.25 in.0.63 cm10 to 50 mL
FQSFB4422180 μm maximum0.125 in. (1/8 in.)0.3 cm0.5 to 15 mL
Catalog number FQSFB4435
Price (MYR)
-
Amplitude:
120 μm maximum
Diameter (English):
0.25 in.
Diameter (Metric):
0.63 cm
Volume Range (Metric):
10 to 50 mL
These Fisher Scientific™ Probes are designed for use with Model 50 and Model 120 Sonic Dismembrators.
  • 5/64 in. Probe microtip for processing 0.2 to 5mL volumes
  • 1/8 in. Probe microtip for processing 0.5 to 15mL volumes
  • 1/4 in. Probe tip for processing 10 to 50mL volumes
Specifications
Product TypeProbe
Amplitude120 μm maximum
Volume Range (Metric)10 to 50 mL
Diameter (English)0.25 in.
Diameter (English) Tip1/4 in.
Diameter (Metric)0.63 cm
Diameter (Metric) Tip6 mm
For Use With (Equipment)Model 120 sonic dismembrator
Packaging Quantity1 probe
Unit SizeEA