Microtip for use with 500 and 700 watt Sonic Dismembrators
Microtip for use with 500 and 700 watt Sonic Dismembrators
Actual product may vary
Microtip for use with 500 and 700 watt Sonic Dismembrators
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume Range (Metric)Diameter (Metric) Tip
FQSFB44205 to 50 mL6 mm
FQSFB44170.2 to 5 mL2 mm
FQSFB44180.5 to 15 mL3 mm
Catalog number FQSFB4420
Price (MYR)
-
Volume Range (Metric):
5 to 50 mL
Diameter (Metric) Tip:
6 mm
These Fisher Scientific™ Probes are designed for use with Model 505 and Model 705 Sonic Dismembrators.
  • Microtip Probes for Model 505/705 Sonic Dismembrator
  • Requires #4220 (1/2 in. Replaceable tip probe)
Specifications
Product TypeMicrotip Probe
Amplitude200 μm maximum
Volume Range (Metric)5 to 50 mL
Diameter (English)0.25 in.
Diameter (English) Tip1/4 in.
Diameter (Metric)0.63 cm
Diameter (Metric) Tip6 mm
For Use With (Equipment)0.5 in. probe (#4220) for Model 505 and 705 sonic dismembrator
IncludesProbe
Packaging Quantity1 probe
Unit SizeEA