Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Actual product may vary
Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
Have Questions?
Catalog NumberDiameter (Metric) TipVolume Range (Metric)Includes
FQSFB421913 mm20 to 250 mLProbe
Catalog number FQSFB4219
Price (MYR)
-
Diameter (Metric) Tip:
13 mm
Volume Range (Metric):
20 to 250 mL
Includes:
Probe
These Fisher Scientific™ Probes are designed for use with 500 and 700 watt Sonic Dismembrators.
  • Available in diameters ranging from 1/16 in. to 1 in.
  • Capacity options of 0.2-5mL to 100-1000mL
  • Replaceable tip, solid and microtip probes
Specifications
Product TypeProbe
Amplitude120 μm maximum
Volume Range (Metric)20 to 250 mL
Diameter (English)0.5 in.
Diameter (English) Tip0.5 in.
Diameter (Metric)1.27 cm
Diameter (Metric) Tip13 mm
For Use With (Application)Ideal for low surface tension liquids
For Use With (Equipment)Model 505/705 Sonic dismembrator
IncludesProbe
Tip StyleSolid
Unit SizeEach