Jack Stand
Jack Stand
Actual product may vary
Jack Stand
Ideal for raising samples up to a stationary probe mounted within the enclosure or on a stand.
Have Questions?
Catalog number FQSFB357
Price (MYR)
-
Fisher Scientific™ Jack Stand is ideal for samples to a stationary probe mounted within the enclosure.
  • Platform that raises and lowers samples under a stationary probe
  • For use with Model 505/705 Sonic Dismembrator.
Specifications
Product TypeJack Stand
Dimensions (L x W)6 x 6 in. platform
For Use With (Application)Ideal to raise samples to a stationary probe mounted in an enclosure
For Use With (Equipment)Model 505/705 sonic dismembrator
Packaging Quantity1 Ea.
Unit SizeEach