Model 120 Sonic Dismembrator
Model 120 Sonic Dismembrator
Actual product may vary
Model 120 Sonic Dismembrator
Programable system ideal for small volume liquid processing. There are 3 probe size options available which allow for processing 0.2-50 mL volumes.
Have Questions?
Catalog NumberElectrical RequirementsIncludes
FQSFB120220220/230 V, 50/60 HzGenerator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
Catalog number FQSFB120220
Price (MYR)
-
Electrical Requirements:
220/230 V, 50/60 Hz
Includes:
Generator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
Use this sonic dismembrator for effective analysis and sample preparation of small sample sizes. Fisher Scientific™ Model 120 Sonic Dismembrator offers both manual and programmable operation for consistent and reproducible results. Processes 0.2 to 50mL.
  • Processes 0.2 to 50mL
  • 120 watts
  • Programmable from 1 second to 10 hours
  • Overload protection
  • Pulse mode (1 second to 59 seconds)
  • Energy Display(Watts and Joules)
  • Available accessories include: 3 Probe sizes for individual samples, Eight tip horn for increased sample throughput, Cup Horn for indirect sonication
  • Lightweight converter for handheld operation
Specifications
Voltage220/230 V
Electrical Requirements220/230 V, 50/60 Hz
Hertz20 kHz (Ultrasonic Frequency)
Wattage120 W
Volume Range (Metric)0.2 to 50 mL
Dimensions (L x W x H)13.5 x 8 x 6 in. (34.29 x 20.32 x 15.24 cm)
Height (English)6 in.
Height (Metric)15.24 cm
Length (English)13.5 in.
Length (Metric)34.29 cm
Width (English)8 in.
Width (Metric)20.32 cm
For Use With (Application)Cell disruption, protein extraction, DNA shearing/ChIP assay
IncludesGenerator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
ModelFB120
Timer1 sec. to 10 hrs.
Unit SizeEach